missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cyclin I Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16357-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. This cyclin shows the highest similarity with cyclin G. The transcript of this gene was found to be expressed constantly during cell cycle progression. The function of this cyclin has not yet been determined.Specifications
cyclin I | |
Polyclonal | |
Unconjugated | |
CCNI | |
CCNI, CYC1, cyclin I, CYI | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10983, 12453, 289500 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14094, Q9Z2V9 | |
Ccni | |
cyclin I Fusion Protein Ag0493 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |