missing translation for 'onlineSavingsMsg'
Learn More

Cyclizine Hydrochloride, LoGiCal

Product Code. 30462000
Change view
Click to view available options
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30462000 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462000 Supplier LoGiCal Supplier No. LGCFOR1265.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Cyclizine Hydrochloride
CAS 303-25-3
Analyt- eller komponentnamn Cyclizine Hydrochloride
Frakt skick Room Temperature
Molekylformel C18 H22 N2 . Cl H
Certifikat och efterlevnad ISO 9001
Synonym Cyclizine hydrochloride, 1-(Diphenylmethyl)-4-methylpiperazine hydrochloride, Piperazine, 1-(diphenylmethyl)-4-methyl-, hydrochloride (1:1), Piperazine, 1-(diphenylmethyl)-4-methyl-, monohydrochloride (8CI,9CI), Marezine hydrochloride, Marzine, N-Benzhydryl-N'-methylpiperazine monohydrochloride
InChI formel InChI=1S/C18H22N2.ClH/c1-19-12-14-20(15-13-19)18(16-8-4-2-5-9-16)17-10-6-3-7-11-17;/h2-11,18H,12-15H2,1H3;1H
Lösningstyp Neat
LEDER Cl.CN1CCN(CC1)C(c2ccccc2)c3ccccc3
IUPAC-namn 1-benzhydryl-4-methylpiperazine;hydrochloride
Industry Type Clinical Testing
Molekylvikt (g/mol) 302.84
Kvantitet 10 mg
Formel vikt 302.155 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.