Läs mer
Cyclizine Hydrochloride, TRC
Produktkod. 30368452
Change view
Kvantitet:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368452 5 g
30365620 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30368452 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9878055G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 303-25-3
Molekylformel C18 H22 N2 . Cl H
Molekylvikt (g/mol) 302.84
Synonym Cyclizine hydrochloride, 1-(Diphenylmethyl)-4-methylpiperazine hydrochloride, Piperazine, 1-(diphenylmethyl)-4-methyl-, hydrochloride (1:1), Piperazine, 1-(diphenylmethyl)-4-methyl-, monohydrochloride (8CI,9CI), Marezine hydrochloride, Marzine, N-Benzhydryl-N'-methylpiperazine monohydrochloride
IUPAC-namn 1-benzhydryl-4-methylpiperazine;hydrochloride
LEDER Cl.CN1CCN(CC1)C(c2ccccc2)c3ccccc3
Specifikationer
Kemiskt namn eller material Cyclizine Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 5 g
InChI formel InChI=1S/C18H22N2.ClH/c1-19-12-14-20(15-13-19)18(16-8-4-2-5-9-16)17-10-6-3-7-11-17;/h2-11,18H,12-15H2,1H3;1H
Formel vikt 302.155
Procent renhet >95%