Läs mer
Cyclizine N-Oxide Dihydrochloride, Mikromol™
Produktkod. 30461264
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30461264 4 x 25 mg
30050492 25 mg
30040809 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30461264 Leverantör Mikromol Leverantörsnummer MM1265.0400254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Cyclizine N-Oxide Dihydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Cyclizine N-Oxide Dihydrochloride
CAS 2725355-17-7
Analyt- eller komponentnamn Cyclizine N-Oxide Dihydrochloride
Frakt skick Room Temperature
Molekylformel C18 H22 N2 O . 2 Cl H
Certifikat och efterlevnad ISO 9001
Synonym Cyclizine N-Oxide Dihydrochloride
InChI formel InChI=1S/C18H22N2O.2ClH/c1-20(21)14-12-19(13-15-20)18(16-8-4-2-5-9-16)17-10-6-3-7-11-17;;/h2-11,18H,12-15H2,1H3;2*1H
Lösningstyp Neat
LEDER Cl.Cl.C[N+]1([O-])CCN(CC1)C(c2ccccc2)c3ccccc3
IUPAC-namn 4-benzhydryl-1-methyl-1-oxidopiperazin-1-ium;dihydrochloride
Molekylvikt (g/mol) 355.3
Kvantitet 4 x 25 mg
Formel vikt 354.1266 g/mol