Läs mer
Cyklo(-gly-phe), TRC
Specifications
Specifications
| Kemiskt namn eller material | Cyclo(-Gly-Phe) |
| CAS | 10125-07-2 |
| Molekylformel | C11H12N2O2 |
| Kvantitet | 250 mg |
| Synonym | (S)-3-(Phenylmethyl)-2,5-piperazinedione, L-3-Benzyl-2,5-piperazinedione, Cyclo(glycyl-L-phenylalanyl), (S)-2-Benzylpiperazine-3,6-dione, (S)-3-Benzyl-2,5-dioxopiperazine, (S)-3-Benzylpiperazine-2,5-dione, Cyclic glycylphenylalanine, Cyclic(glycylphenylalanyl), Cyclo(Gly-L-Phe), Cyclo(L-phenylalanylglycine), Cyclo(L-phenylalanylglycyl), Cyclo-glycyl-L-phenylalanine |
| InChI formel | InChI=1S/C11H12N2O2/c14-10-7-12-11(15)9(13-10)6-8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,12,15)(H,13,14)/t9-/m0/s1 |
| LEDER | O=C1CNC(=O)[C@H](Cc2ccccc2)N1 |
| IUPAC-namn | (3S)-3-benzylpiperazin-2,5-dione |
| Molekylvikt (g/mol) | 204.23 |
| Formel vikt | 204.0899 |