Läs mer
Cyclo(-Gly-Tyr), TRC
Produktkod. 30362794
Change view
Kvantitet:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30362794 250 mg
30356363 1 g
30352808 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30362794 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC987420250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5845-66-9
Molekylformel C11H12N2O3
Molekylvikt (g/mol) 220.22
Synonym L-3-(p-Hydroxybenzyl)-2,5-piperazinedione, (S)-3-[(4-Hydroxyphenyl)methyl]-2,5-piperazinedione, 3-(S)-(4-Hydroxybenzyl)piperazine-2,5-dione, Cyclic(glycyl-L-tyrosine), Cyclic(glycyltyrosyl), Cyclo(Gly-L-Tyr), Cyclo(L-tyrosylglycine)
IUPAC-namn (3S)-3-[(4-hydroxyphenyl)methyl]piperazine-2,5-dione
LEDER O=C(NC1)[C@@H](NC1=O)CC2=CC=C(O)C=C2
Specifikationer
Kemiskt namn eller material Cyclo(-Gly-Tyr)
InChI formel InChI=1S/C11H12N2O3/c14-8-3-1-7(2-4-8)5-9-11(16)12-6-10(15)13-9/h1-4,9,14H,5-6H2,(H,12,16)(H,13,15)/t9-/m0/s1
Kvantitet 250 mg
Formel vikt 220.0848