Läs mer
Cyclo(L-Ala-L-Gly), TRC
Produktkod. 30360248
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30360248 100 mg
1 options
Produktkod. 30360248 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC989770100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Cyclo(L-Ala-L-Gly)
CAS 4526-77-6
Molekylformel C5H8N2O2
Kvantitet 100 mg
Synonym (S)-3-Methyl-2,5-piperazinedione, L-3-Methyl-2,5-piperazinedione, Cyclo(L-alanylglycyl), Alanine, cyclic anhydride with glycine, Cyclo(Ala-Gly), Cyclo(Gly-L-Ala), Cyclo(alanylglycine), Cyclo(glycylalanine), N-Glycylalanine cyclic anhydride
InChI formel InChI=1S/C5H8N2O2/c1-3-5(9)6-2-4(8)7-3/h3H,2H2,1H3,(H,6,9)(H,7,8)/t3-/m0/s1
LEDER C[C@@H]1NC(=O)CNC1=O
IUPAC-namn (3S)-3-methylpiperazine-2,5-dione
Molekylvikt (g/mol) 128.13
Formel vikt 128.0586