Läs mer
Cyclo(L-prolyl-L-phenylalanyl), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Cyclo(L-prolyl-L-phenylalanyl) |
| CAS | 3705-26-8 |
| Molekylformel | C14H16N2O2 |
| Kvantitet | 250 mg |
| Synonym | (3S,8aS)-3-Benzylhexahydro-pyrrolo[1,2-a]pyrazine-1,4-dione, (3S-trans)-Hexahydro-3-(phenylmethyl)-pyrrolo[1,2-a]pyrazine-1,4-dione, (3S,8aS)-Hexahydro-3-(phenylmethyl)pyrrolo[1,2-a]pyrazine-1,4-dione, Cyclo(L-Phe-L-Pro), Cyclo(L-Pro-L-Phe), Cyclo(L-phenylalanyl-L-prolyl-), Cyclo(Phe-Pro), L-Phenylalanyl-L-proline Lactam, L-Prolyl-L-phenylalanine Diketopiperazine, Maculosin 2, Maculosine 2, SMFEI 02 |
| InChI formel | InChI=1S/C14H16N2O2/c17-13-12-7-4-8-16(12)14(18)11(15-13)9-10-5-2-1-3-6-10/h1-3,5-6,11-12H,4,7-9H2,(H,15,17)/t11-,12-/m0/s1 |
| LEDER | O=C1N[C@@H](Cc2ccccc2)C(=O)N3CCC[C@@H]13 |
| IUPAC-namn | (3S,8aS)-3-benzyl-2,3,6,7,8,8a-hexahydropyrrolo[1,2-a]pyrazine-1,4-dione |
| Molekylvikt (g/mol) | 244.29 |
| Formel vikt | 244.1212 |