Läs mer
Cyclo(L-prolyl-L-valyl), TRC
Produktkod. 30359988
Change view
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30359988 25 mg
30352684 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30359988 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC98982525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2854-40-2
Molekylformel C10H16N2O2
Molekylvikt (g/mol) 196.25
Synonym (3S-trans)-Hexahydro-3-(1-methylethyl)-pyrrolo[1,2-a]pyrazine-1,4-dione
IUPAC-namn (3S,8aS)-3-isopropylhexahydropyrrolo[1,2-a]pyrazine-1,4-dione
LEDER O=C([C@@H](N1)C(C)C)N(CCC2)[C@]2([H])C1=O
Specifikationer
Kemiskt namn eller material Cyclo(L-prolyl-L-valyl)
Kvantitet 25 mg
InChI formel InChI=1S/C10H16N2O2/c1-6(2)8-10(14)12-5-3-4-7(12)9(13)11-8/h6-8H,3-5H2,1-2H3,(H,11,13)/t7-,8-/m0/s1
Formel vikt 196.12