missing translation for 'onlineSavingsMsg'
Läs mer

Cyclo(Leu-Gly), TRC

Product Code. 30357244
Change view
Click to view available options
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30357244 50 mg
30362201 250 mg
30363453 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357244 Supplier Toronto Research Chemicals Supplier No. TRCC98448550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5845-67-0
Molekylformel C8H14N2O2
Molekylvikt (g/mol) 170.21
Synonym Cyclo(Gly-L-Leu), (S)-3-Isobutylpiperazine-2,5-dione, (3S)-3-(2-Methylpropyl)piperazine-2,5-dione, L-3-Isobutyl-2,5-piperazinedione, Cyclo(glycyl-L-leucyl), Cyclo(L-leucylglycine), Cyclo(L-leucylglycyl), L-Leucylglycine Diketopiperazine
IUPAC-namn (3S)-3-(2-methylpropyl)piperazine-2,5-dione
LEDER CC(C)C[C@@H]1NC(=O)CNC1=O

Specifications

Kemiskt namn eller material Cyclo(Leu-Gly)
InChI formel InChI=1S/C8H14N2O2/c1-5(2)3-6-8(12)9-4-7(11)10-6/h5-6H,3-4H2,1-2H3,(H,9,12)(H,10,11)/t6-/m0/s1
Kvantitet 50 mg
Formel vikt 170.1055
Frakt skick Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.