Läs mer
Cyclofenil, TRC
Produktkod. 30007446
Change view
Kvantitet:
10 mg
50 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007446 10 mg
30006490 50 mg
30005629 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007446 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC98783510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2624-43-3
Molekylformel C23 H24 O4
Molekylvikt (g/mol) 364.43
Synonym Phenol, 4,4'-(cyclohexylidenemethylene)bis-, diacetate (9CI), Phenol, 4-[[4-(acetyloxy)phenyl]cyclohexylidenemethyl]-, acetate, p-Cresol, α-cyclohexylidene-α-(p-hydroxyphenyl)-, diacetate (7CI,8CI), 4,4'-(Cyclohexylidenemethylene)diphenol diacetate ester, Bis(p-acetoxyphenyl)cyclohexylidenemethane, Bis(p-hydroxyphenyl)cyclohexyldienemethane diacetate, Cyclofenil, Cyclofenyl, Cyclopenil, Cyclophenil, Cyclophenyl, F 6066, Fertodur, H 3452, ICI 48213, NSC 86464, Neoclym, Oginex, Ondogyne, Ondonid, Rehibin, Sanocrisin, Sexadieno, Sexovar, Sexovid, α-(p-Hydroxyphenyl)-α-cyclohexylidene-p-cresol diacetate, α-Cyclohexylidene-α-(p-hydroxyphenyl)-p-cresol diacetate, Phenol, 4,4'-(cyclohexylidenemethylene)bis-, 1,1'-diacetate
IUPAC-namn [4-[(4-acetyloxyphenyl)-cyclohexylidenemethyl]phenyl] acetate
LEDER CC(=O)Oc1ccc(cc1)C(=C2CCCCC2)c3ccc(OC(=O)C)cc3
Specifikationer
Kemiskt namn eller material Cyclofenil
Kvantitet 10 mg
InChI formel InChI=1S/C23H24O4/c1-16(24)26-21-12-8-19(9-13-21)23(18-6-4-3-5-7-18)20-10-14-22(15-11-20)27-17(2)25/h8-15H,3-7H2,1-2H3
Formel vikt 364.1675