Läs mer
Cyclohexanecarbohydrazide, TRC
Produktkod. 30363760
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30363760 50 mg
30354914 100 mg
30359916 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30363760 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC98730350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 38941-47-8
Molekylformel C7H14N2O
Molekylvikt (g/mol) 142.19
IUPAC-namn cyclohexanecarbohydrazide
LEDER NNC(=O)C1CCCCC1
Specifikationer
Kemiskt namn eller material Cyclohexanecarbohydrazide
Kvantitet 50 mg
InChI formel InChI=1S/C7H14N2O/c8-9-7(10)6-4-2-1-3-5-6/h6H,1-5,8H2,(H,9,10)
Formel vikt 142.1106