Läs mer
Cyclohexyl Butyl Phthalate, TRC
Produktkod. 30365483
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30365483 500 mg
1 options
Produktkod. 30365483 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC988140500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Butyl cyclohexyl phthalate
CAS 84-64-0
Molekylformel C18 H24 O4
Kvantitet 500 mg
Synonym 1,2-Benzenedicarboxylic acid, butyl cyclohexyl ester (9CI), Phthalic acid, butyl cyclohexyl ester (6CI,7CI,8CI), Butyl cyclohexyl phthalate, Cyclohexyl butyl phthalate, Elastex 50B, NSC 69994
InChI formel InChI=1S/C18H24O4/c1-2-3-13-21-17(19)15-11-7-8-12-16(15)18(20)22-14-9-5-4-6-10-14/h7-8,11-12,14H,2-6,9-10,13H2,1H3
LEDER CCCCOC(=O)c1ccccc1C(=O)OC2CCCCC2
IUPAC-namn 1-O-butyl 2-O-cyclohexyl benzene-1,2-dicarboxylate
Molekylvikt (g/mol) 304.38
Formel vikt 304.1675