Learn More
Cyclohexylacetylene, 98%
Product Code. 15450208
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450208 25 g 25g
15440208 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15450208 Supplier Thermo Scientific Alfa Aesar Supplier No. H53418.14
 Restricted Item
Beräknad leverans: 30-12-2026
Add to basket
CAS: 931-48-6 | C8H12 | 108.184 g/mol
Cyclohexylacetylene is used in the preparation of hydrido-vinylidene complexes of osmium, which finds vital intermediates in several homogeneous and heterogeneous catalytic reactions such as alkene olimerization, polymerization and Fischer-Tropsch synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclohexylacetylene is used in the preparation of hydrido-vinylidene complexes of osmium, which finds vital intermediates in several homogeneous and heterogeneous catalytic reactions such as alkene olimerization, polymerization and Fischer-Tropsch synthesis.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 931-48-6
Molecular Formula C8H12
Molecular Weight (g/mol) 108.184
MDL Number MFCD00001513
InChI Key SSDZYLQUYMOSAK-UHFFFAOYSA-N
Synonym cyclohexylacetylene, cyclohexylethyne, cyclohexane, ethynyl, cyclohexyl acetylene, ethynyl-cyclohexane, cyclohexane, ethynyl-6ci, 7ci, 8ci, 9ci
PubChem CID 70263
IUPAC Name ethynylcyclohexane
SMILES C#CC1CCCCC1
Specifications
Density 0.828
Boiling Point 130°C to 132°C
Flash Point 18°C (64°F)
Refractive Index 1.454
Quantity 25 g
UN Number UN3295
Beilstein 1815535
Solubility Information Immiscible with water.
Formula Weight 108.18
Percent Purity 98%
Chemical Name or Material Cyclohexylacetylene