Learn More
Cyclohexylamine hydrochloride, 99%, Thermo Scientific Chemicals
Catalog Number p-96806
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
4998-76-9
C6H14ClN
135.64
MFCD00050540
ZJUGSKJHHWASAF-UHFFFAOYSA-N
cyclohexylamine hydrochloride, cyclohexanamine hydrochloride, cyclohexanamine, hydrochloride, aminocyclohexane hydrochloride, cyclohexylamine.hcl, unii-tn6vyf8thc, dsstox_cid_361, cyclohexanamine, hydrochloride 1:1, dsstox_rid_75539, dsstox_gsid_20361
2723770
cyclohexanamine;hydrochloride
[Cl-].[NH3+]C1CCCCC1
4998-76-9
C6H14ClN
135.64
MFCD00050540
ZJUGSKJHHWASAF-UHFFFAOYSA-N
cyclohexylamine hydrochloride, cyclohexanamine hydrochloride, cyclohexanamine, hydrochloride, aminocyclohexane hydrochloride, cyclohexylamine.hcl, unii-tn6vyf8thc, dsstox_cid_361, cyclohexanamine, hydrochloride 1:1, dsstox_rid_75539, dsstox_gsid_20361
2723770
cyclohexanamine;hydrochloride
[Cl-].[NH3+]C1CCCCC1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 207.0°C
Color White to Beige
Boiling Point 306.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 100 g
Solubility Information Solubility in water: 83 g/100mL water (17°C). Other solubilities: soluble in most common organic solvents, practically insoluble in ethers
Formula Weight 135.63
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Cyclohexylamine hydrochloride, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclohexylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361f-Suspected of damaging fertility.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.