Läs mer
Cyclopentamine Hydrochloride, LoGiCal
Produktkod. 30464187
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30464187 5 mg
1 options
Produktkod. 30464187 Leverantör LoGiCal Leverantörsnummer LGCFOR3027.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Cyclopentamine Hydrochloride
CAS 538-02-3
Analyt- eller komponentnamn Cyclopentamine Hydrochloride
Frakt skick Room Temperature
Molekylformel C9 H19 N . Cl H
Certifikat och efterlevnad ISO 9001
Synonym Cyclopentaneethanamine, N,α-dimethyl-, hydrochloride, Cyclopentamine hydrochloride, Cyclopentaneethanamine, N,α-dimethyl-, hydrochloride, Cyclopentaneethylamine, N,α-dimethyl-, hydrochloride, (±)-Cyclopentamine hydrochloride, 1-Cyclopentyl-2-methylaminopropane hydrochloride, Clopane hydrochloride, N,α-Dimethylcyclopentaneethylamine hydrochloride
InChI formel InChI=1S/C9H19N.ClH/c1-8(10-2)7-9-5-3-4-6-9;/h8-10H,3-7H2,1-2H3;1H
Lösningstyp Neat
LEDER Cl.CNC(C)CC1CCCC1
IUPAC-namn 1-cyclopentyl-N-methylpropan-2-amine;hydrochloride
Industry Type Clinical Testing
Molekylvikt (g/mol) 177.71
Kvantitet 5 mg
Formel vikt 177.1284 g/mol