Läs mer
Cyclopentamine Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Cyclopentamine Hydrochloride |
| CAS | 538-02-3 |
| Molekylformel | C9 H19 N . Cl H |
| Kvantitet | 25 mg |
| Synonym | Cyclopentaneethanamine, N,α-dimethyl-, hydrochloride, Cyclopentamine hydrochloride, Cyclopentaneethanamine, N,α-dimethyl-, hydrochloride, Cyclopentaneethylamine, N,α-dimethyl-, hydrochloride, (±)-Cyclopentamine hydrochloride, 1-Cyclopentyl-2-methylaminopropane hydrochloride, Clopane hydrochloride, N,α-Dimethylcyclopentaneethylamine hydrochloride |
| InChI formel | InChI=1S/C9H19N.ClH/c1-8(10-2)7-9-5-3-4-6-9;/h8-10H,3-7H2,1-2H3;1H |
| LEDER | Cl.CNC(C)CC1CCCC1 |
| IUPAC-namn | 1-cyclopentyl-N-methylpropan-2-amine;hydrochloride |
| Molekylvikt (g/mol) | 177.71 |
| Formel vikt | 177.1284 |