missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Cyclopentolate N-Oxide Hydrochloride, Mikromol™
Beskrivning
Cyclopentolate N-Oxide Hydrochloride
Specifikationer
Specifikationer
| Kemiskt namn eller material | Cyclopentolate N-Oxide Hydrochloride |
| CAS | 2724726-92-3 |
| Analyt- eller komponentnamn | Cyclopentolate N-Oxide Hydrochloride |
| Frakt skick | Ice pack (-20°C) |
| Molekylformel | C17 H25 N O4 . Cl H |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | Cyclopentolate N-Oxide Hydrochloride |
| InChI formel | InChI=1S/C17H25NO4.ClH/c1-18(2,21)12-13-22-16(19)15(14-8-4-3-5-9-14)17(20)10-6-7-11-17;/h3-5,8-9,15,20H,6-7,10-13H2,1-2H3;1H |
| LEDER | Cl.C[N+](C)([O-])CCOC(=O)C(c1ccccc1)C2(O)CCCC2 |
| IUPAC-namn | 2-[2-(1-hydroxycyclopentyl)-2-phenylacetyl]oxy-N,N-dimethylethanamine oxide;hydrochloride |
| Visa mer |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?