Learn More
Cyclopentyl isocyanate, 97%, Thermo Scientific Chemicals
Catalog Number p-3771230
Click to view available options
Quantity:
1 g
5 g
4747-71-1
C6H9NO
111.14
MFCD02093695
CZALJDQHONFVFU-UHFFFAOYSA-N
cyclopentyl isocyanate, cyclopentylisocyanate, cyclopentane, isocyanato, isocyanic acid cyclopentyl ester, isocyanato cyclopentane, isocyanato-cyclopentane, acmc-1aspa, ksc497s6f
643500
isocyanatocyclopentane
C1CCC(C1)N=C=O
4747-71-1
C6H9NO
111.14
MFCD02093695
CZALJDQHONFVFU-UHFFFAOYSA-N
cyclopentyl isocyanate, cyclopentylisocyanate, cyclopentane, isocyanato, isocyanic acid cyclopentyl ester, isocyanato cyclopentane, isocyanato-cyclopentane, acmc-1aspa, ksc497s6f
643500
isocyanatocyclopentane
C1CCC(C1)N=C=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 0.9860g/mL
Boiling Point 147.0°C
Flash Point 36°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C5H9NCO
Quantity 1 g
Specific Gravity 0.986
Formula Weight 111.14
Percent Purity 97%
Chemical Name or Material Cyclopentyl isocyanate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclopentyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .