Läs mer
Cyclopentylmethylamine, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Cyclopentylmethylamine |
| CAS | 2439-56-7 |
| Molekylformel | C6H13N |
| Kvantitet | 250 mg |
| Synonym | N-Methylcyclopentanamine, (Methylamino)cyclopentane, Methyl(cyclopentyl)amine, N-Cyclopentyl-N-methylamine, N-Methylcyclopentylamine |
| InChI formel | InChI=1S/C6H13N/c1-7-6-4-2-3-5-6/h6-7H,2-5H2,1H3 |
| LEDER | CNC1CCCC1 |
| IUPAC-namn | N-methylcyclopentanamine |
| Molekylvikt (g/mol) | 99.17 |
| Formel vikt | 99.1048 |