Läs mer
Cyclopentynafil, TRC
Produktkod. 30352832
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30352832 250 mg
30349556 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30352832 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC989015250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1173706-34-7
Molekylformel C26 H36 N6 O4 S
Molekylvikt (g/mol) 528.67
Synonym 5-(5-((4-Cyclopentylpiperazin-1-yl)sulfonyl)-2-ethoxyphenyl)-1-methyl-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-7(6H)-one, 5-[5-[(4-Cyclopentyl-1-piperazinyl)sulfonyl]-2-ethoxyphenyl]-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one, Cyclopentynafil
IUPAC-namn 5-[5-(4-cyclopentylpiperazin-1-yl)sulfonyl-2-ethoxyphenyl]-1-methyl-3-propyl-4H-pyrazolo[4,3-d]pyrimidin-7-one
LEDER CCCc1nn(C)c2C(=O)N=C(Nc12)c3cc(ccc3OCC)S(=O)(=O)N4CCN(CC4)C5CCCC5
Specifikationer
Kemiskt namn eller material Cyclopentynafil
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C26H36N6O4S/c1-4-8-21-23-24(30(3)29-21)26(33)28-25(27-23)20-17-19(11-12-22(20)36-5-2)37(34,35)32-15-13-31(14-16-32)18-9-6-7-10-18/h11-12,17-18H,4-10,13-16H2,1-3H3,(H,27,28,33)
Formel vikt 528.2519
Procent renhet >95%