Learn More
Cyclopropyl-1-d1 Methyl-d3 Ketone, CDN
Product Code. 30317188
Change view
Click to view available options
Kvantitet:
1 g
Product Code. Kvantitet
30317188 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30317188 Supplier CDN Supplier No. CDND50481G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 1-Cyclopropylethanone-1',2,2,2-D4
CAS 95249-93-7
Molekylformel C5 D4 H4 O
Kvantitet 1 g
Synonym Ethanone-2,2,2-d3, 1-(cyclopropyl-1-d)- (9 CI), 1-(Cyclopropyl-1-d)ethanone-2,2,2-d3, Cyclopropyl-1-d1 Methyl-d3 Ketone, 1-Cyclopropylethanone-D4, 1-Cyclopropylethanone-1',2,2,2-D4
InChI formel InChI=1 S/C5H8O/c1-4(6)5-2-3-5/h5H,2-3H2,1H3/i1D3,5 D
LEDER [2 H]C([2 H])([2 H])C(=O)C1([2 H])CC1
IUPAC-namn 2,2,2-trideuterio-1-(1-deuteriocyclopropyl)ethanone
Molekylvikt (g/mol) 88.1411
Formel vikt 88.0826 g/mol
Procent renhet 98 atom % D, min 98% Chemical Purity