Läs mer
Cyclopropyl 2-Fluorophenyl Diketone, TRC
Produktkod. 30134863
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134863 100 mg
17941075 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134863 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC988705100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391054-37-7
Molekylformel C11 H9 F O2
Molekylvikt (g/mol) 192.19
Synonym 1-Cyclopropyl-2-(2-fluorophenyl)ethane-1,2-dione
IUPAC-namn 1-cyclopropyl-2-(2-fluorophenyl)ethane-1,2-dione
LEDER Fc1ccccc1C(=O)C(=O)C2CC2
Specifikationer
Kemiskt namn eller material 1-Cyclopropyl-2-(2-fluorophenyl)ethane-1,2-dione
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C11H9FO2/c12-9-4-2-1-3-8(9)11(14)10(13)7-5-6-7/h1-4,7H,5-6H2
Formel vikt 192.0587
Procent renhet >95%