missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylcarboxamidine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number p-7024920
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
57297-29-7
C4H9ClN2
120.58
MFCD00053010
JRYOZJIRAVZGMV-UHFFFAOYSA-N
cyclopropanecarboximidamide hydrochloride, cyclopropane-1-carboximidamide hydrochloride, cyclopropanecarboxamidine hydrochloride, cyclopropylcarbamidine hydrochloride, cyclopropanecarboxamidine hcl, cyclopropyl carbamidine hydrochloride, clcyopropylcarbamidine hydrochloride, cyclopropylcarboxamidine hydrochloride, cyclopropyl carbamidine hcl, cyclopropanecarboximidamide, monohydrochloride
2781916
[H+].[Cl-].NC(=N)C1CC1
This item is not returnable. View return policy
57297-29-7
C4H9ClN2
120.58
MFCD00053010
JRYOZJIRAVZGMV-UHFFFAOYSA-N
cyclopropanecarboximidamide hydrochloride, cyclopropane-1-carboximidamide hydrochloride, cyclopropanecarboxamidine hydrochloride, cyclopropylcarbamidine hydrochloride, cyclopropanecarboxamidine hcl, cyclopropyl carbamidine hydrochloride, clcyopropylcarbamidine hydrochloride, cyclopropylcarboxamidine hydrochloride, cyclopropyl carbamidine hcl, cyclopropanecarboximidamide, monohydrochloride
2781916
[H+].[Cl-].NC(=N)C1CC1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 122°C to 123°C
Quantity 1 g
Beilstein 4507255
Sensitivity Hygroscopic
IUPAC Name cyclopropanecarboximidamide;hydrochloride
Formula Weight 120.59
Percent Purity 97%
Chemical Name or Material Cyclopropylcarboxamidine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'