missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropylmagnesium bromide, 0.5M in MeTHF, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mL
100 mL
Description
Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).
Solubility
Reacts with water.
Notes
Air and Moisture Sensitive, store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).
Solubility
Reacts with water.
Notes
Air and Moisture Sensitive, store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Quantity | 50 mL |
UN Number | UN3399 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Reacts with water. |
IUPAC Name | magnesium;cyclopropane;bromide |
Formula Weight | 145.28 |
Concentration or Composition (by Analyte or Components) | 0.5M in MeTHF |
Chemical Name or Material | Cyclopropylmagnesium bromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'