missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropyltriphenylphosphonium bromide, 98%, Thermo Scientific Chemicals
330.56 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
It is used for electrolytic polarization of Zn, has potential antitumor activity, reactant for synthesis of cycloporpylidenemethylarylethynynes, keto halides for sequential azidation and intramolecular Schmidt reactions, a reactant for hydroalkynylation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used for electrolytic polarization of Zn, has potential antitumor activity, reactant for synthesis of cycloporpylidenemethylarylethynynes, keto halides for sequential azidation and intramolecular Schmidt reactions, a reactant for hydroalkynylation.
Solubility
Soluble in water.
Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
It is used for electrolytic polarization of Zn, has potential antitumor activity, reactant for synthesis of cycloporpylidenemethylarylethynynes, keto halides for sequential azidation and intramolecular Schmidt reactions, a reactant for hydroalkynylation.
Solubility
Soluble in water.
Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
Specifications
Melting Point | 183°C to 186°C |
Quantity | 5 g |
Beilstein | 6413483 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 383.27 |
Percent Purity | 98% |
Chemical Name or Material | Cyclopropyltriphenylphosphonium bromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'