Läs mer
Cycloserine Diketopiperazine, TRC
Produktkod. 30353477
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30353477 25 mg
30364312 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30353477 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC98881025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1204-99-5
Molekylformel C6H12N4O4
Molekylvikt (g/mol) 204.18
Synonym 3,6-Bis[(aminooxy)methyl]-2,5-piperazinedione, 2,5-Bis(aminohydroxymethyl)-2,6- diketopiperazine, 2,5-Bis(aminooxymethyl)-3,6-dioxopiperazine, Cycloserine Diketopiperazine, NSC 119130
IUPAC-namn 3,6-bis(aminooxymethyl)piperazine-2,5-dione
LEDER C(C1C(=O)NC(C(=O)N1)CON)ON
Specifikationer
Kemiskt namn eller material Cycloserine Diketopiperazine
InChI formel InChI=1S/C6H12N4O4/c7-13-1-3-5(11)10-4(2-14-8)6(12)9-3/h3-4H,1-2,7-8H2,(H,9,12)(H,10,11)
Kvantitet 25 mg
Formel vikt 204.0859