Läs mer
Cyclosporin G, TRC
Produktkod. 30137386
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137386 100 mg
30009999 10 mg
30010899 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137386 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC990143100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 74436-00-3
Synonym Ciclosporin G∣7-L-Norvaline-cyclosporin A∣Cyclosporin G∣1,4,7,10,13,16,19,22,25,28,31-Undecaazacyclotritriacontane, cyclic peptide deriv.∣7-L-Norvaline cyclosporin A∣Cyclo[(2S,3R,4R,6E)-3-hydroxy-4-methyl-2-(methylamino)-6-octenoyl-L-norvalyl-N-methylglycyl-N-methyl-L-leucyl-L-valyl-N-methyl-L-leucyl-L-alanyl-D-alanyl-N-methyl-L-leucyl-N-methyl-L-leucyl-N-methyl-L-valyl]∣Cyclo[L-alanyl-D-alanyl-N-methyl-L-leucyl-N-methyl-L-leucyl-N-methyl-L-valyl-(3R,4R,6E)-6,7-didehydro-3-hydroxy-N,4-dimethyl-L-2-aminooctanoyl-L-norvalyl-N-methylglycyl-N-methyl-L-leucyl-L-valyl-N-methyl-L-leucyl]∣Cyclosporine G∣Geclosporin∣OG 37-325∣SDZ 37-325
Specifikationer
Renhet >95% (HPLC)
Formatera Neat
Molekylvikt (g/mol) 1216.64