Läs mer
Cycrimine Hydrochloride, TRC
Produktkod. 30361184
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30361184 50 mg
30359005 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30361184 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC99145050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 126-02-3
Molekylformel C19 H29 N O . Cl H
Molekylvikt (g/mol) 323.9
Synonym 1-Phenyl-1-cyclopentyl-3-piperidino-1-propanol hydrochloride, Compound 8958, Cycrimine hydrochloride, NSC 169452, Pagitane hydrochloride, 1-Piperidinepropanol, alpha-cyclopentyl-alpha-phenyl-, hydrochloride (1:1), 1-Piperidinepropanol, alpha-cyclopentyl-alpha-phenyl-, hydrochloride, alpha-Cyclopentyl-alpha-phenyl-1-piperidinepropanol hydrochloride
IUPAC-namn 1-cyclopentyl-1-phenyl-3-piperidin-1-ylpropan-1-ol;hydrochloride
LEDER Cl.OC(CCN1CCCCC1)(C2CCCC2)c3ccccc3
Specifikationer
Kemiskt namn eller material Cycrimine Hydrochloride
Kvantitet 50 mg
InChI formel InChI=1S/C19H29NO.ClH/c21-19(18-11-5-6-12-18,17-9-3-1-4-10-17)13-16-20-14-7-2-8-15-20;/h1,3-4,9-10,18,21H,2,5-8,11-16H2;1H
Formel vikt 323.2016