Läs mer
λ-Cyhalothrin, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | lambda-Cyhalothrin |
| CAS | 91465-08-6 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C23 H19 Cl F3 N O3 |
| Kvantitet | 500 mg |
| Synonym | Cyclopropanecarboxylic acid, 3-(2-chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethyl-, cyano(3-phenoxyphenyl)methyl ester, [1α(S*),3α(Z)]-(±)-, Cyclopropanecarboxylic acid, 3-[(1Z)-2-chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethyl-, (R)-cyano(3-phenoxyphenyl)methyl ester, (1S,3S)-rel- (9CI), A 12871A, Ambush Super, Cyclopropanecarboxylic acid, 3-[(1Z)-2-chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethyl-, (1R)-cyano(3-phenoxyphenyl)methyl ester, (1S,3S)-, Cyhalothrin K, Demand, Demand Pestab, Grenade ER, Hallmark, ICI-A 0321, ICON Maxx, Icon, Karate, Karate 1E, Karate 2.5EC, Karate K, Karate King, Karate Z, Karate Zeon, Karate Zeon 050CS, Karate Zeon 100CS, Kung Fu, Lamdex, Matador, Matador (insecticide), Molniya, NUP 05077, PP 321, REEVA 5, Scimitar, Spectracide Trazicide, Spectracide Triazicide Soil and Turf Insect Killer, Taiga Z, Warrior, Warrior II, Warrior T, Warrior Zeon, lambda-Cyhalothrin, λ-Cyhalothrin |
| InChI formel | InChI=1S/C23H19ClF3NO3/c1-22(2)17(12-19(24)23(25,26)27)20(22)21(29)31-18(13-28)14-7-6-10-16(11-14)30-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3/b19-12-/t17-,18+,20-/m1/s1 |
| LEDER | CC1(C)[C@H](\C=C(/Cl)\C(F)(F)F)[C@@H]1C(=O)O[C@@H](C#N)c2cccc(Oc3ccccc3)c2 |
| IUPAC-namn | [(R)-cyano-(3-phenoxyphenyl)methyl] (1S,3S)-3-[(Z)-2-chloro-3,3,3-trifluoroprop-1-enyl]-2,2-dimethylcyclopropane-1-carboxylate |
| Molekylvikt (g/mol) | 449.85 |
| Visa mer |