Learn More
Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels
For the safe, easy and accurate transfer of fluids in the laboratory. Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ B0496501
Additional Details : Weight : 0.18000kg
Product Code. 10244650
  • 1400.00 SEK / Each
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
Description
Description
  • With inscription label
  • With NS socket and cone
  • With scale (approx. volume/mL)
  • With pressure equalizing tube
  • With screwthread retaining device
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
100 mL
Duran Glass, Borosilicate Glass
Pressure Equalization Tube at the Side