Learn More
Lenz Laborglasinstrumente™ Pressure Equalizing Dropping Funnels, with PTFE Plug
Product Code. 12556229
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Joint Number:
14/23 NS
29/32 NS
Includes:
Pressure Equalizing Tube
w/o Pressure Equalizing Tube
Unit Size:
Each
Pack of 2
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Joint Number Includes Unit Size
12556229 50 mL 29/32 NS Pressure Equalizing Tube Pack of 2
30564740 1000 mL 29/32 NS Pressure Equalizing Tube Each
10589041 1000 mL 29/32 NS Pressure Equalizing Tube Pack of 2
10047810 1000 mL 29/32 NS w/o Pressure Equalizing Tube Pack of 2
10635041 100 mL 14/23 NS Pressure Equalizing Tube Pack of 2
10343001 100 mL 29/32 NS w/o Pressure Equalizing Tube Pack of 2
10500012 250 mL 29/32 NS w/o Pressure Equalizing Tube Pack of 2
10519241 25 mL 14/23 NS w/o Pressure Equalizing Tube Pack of 2
10740651 250 mL 29/32 NS Pressure Equalizing Tube Pack of 2
10149650 500 mL 29/32 NS w/o Pressure Equalizing Tube Pack of 2
10707711 25 mL 14/23 NS Pressure Equalizing Tube Pack of 2
12586229 50 mL 14/23 NS Pressure Equalizing Tube Pack of 2
10590002 50 mL 29/32 NS w/o Pressure Equalizing Tube Pack of 2
10792132 500 mL 29/32 NS Pressure Equalizing Tube Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 12556229 Supplier Lenz Laborglasinstrumente™ Supplier No. 04372814
  • ONLINE EXCLUSIVE PRICE
    2260.00 SEK
    1301.06 SEK / Paket med 2
    Save 958.94 SEK
    42% Av
Beräknad leverans: 03-12-2026
Add to basket
Add to basket
Useful for transferring fluids and handling air-sensitive reagents in a sealed, inert-gas environment. Lenz Laborglasinstrumente™ Pressure Equalizing Dropping Funnels are manufactured from durable DURAN™ borosilicate glass.
  • With scale (approx. volume/mL)
  • With pressure equalizing tube
  • With screwthread retaining device
  • Cylindrical shape with NS socket and cone
  • Corrosion-free and resistant to thermal shock
Specifications
Capacity (Metric) 50 mL
Material Borosilicate Glass
Graduated Yes
Includes Pressure Equalizing Tube
Joint Number 29/32 NS
Product Type Dropping Funnel