Learn More
BRAND™ PTFE Magnetic Stirring Bars with Pivot Ring, Cylindrical Shape
Manufactured from PTFE. BRAND™ PTFE Magnetic Stirring Bars with Pivot Ring, Cylindrical Shape are for use with vessels that have slightly uneven bottoms. Pivot ring reduces the contact surface and enables the stirrer to adopt the optimum stirring position.
Brand:  BRAND™ 137415
339.42 SEK valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 0.01980kg
 View more versions of this product
Product Code. 11853960
  • 427.00 SEK / Pack of 10
Estimated Shipment: 07-05-2024
to see stock.
Add to basket
Description
Description
  • Manufactured from PTFE.
  • Cylindrical shape.
  • For use with vessels that have slightly uneven bottoms.
  • Pivot ring reduces the contact surface and enables the stirrer to adopt the optimum stirring position.
  • Manufactured from PTFE.
  • Cylindrical shape.
  • For use with vessels that have slightly uneven bottoms.
  • Pivot ring reduces the contact surface and enables the stirrer to adopt the optimum stirring position.
Specifications
Specifications
8 mm
PTFE
20 mm
10/Pk.
For vessels with slightly uneven bottoms
Cylinder