Läs mer
Tocris Bioscience™ CYM 5442 hydrochloride
Beskrivning
CYM 5442 hydrochloride is a potent and selective S1P1 agonist in vitro (EC50 = 1.35 nM). Activates S1P1-mediated p42/p44 MAPK phosphorylation in CHO-K1 cells transfected with S1P1. Induces acute lymphopenia in mice. Brain penetrant.
Specifikationer
Specifikationer
| CAS | 1783987-80-3 |
| Kvantitet | 50 mg |
| Produkttyp | CYM 5442 hydrochloride |
| Renhet | >98% |
| Molekylvikt (g/mol) | 445.94 |
| Inhibitorer | CYM 5442 hydrochloride |
For Research Use Only