Learn More
Abnova™ CYP19A1 (Human) Recombinant Protein
Human CYP19A1 full-length ORF ( AAH22896, 1 a.a. - 181 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001588-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MVLEMLNPIHYNITSIVPEAMPAATMPVLLLTGLFLLVWNYEGTSSIPGPGYCMGIGPLISHGRFLWMGIGSACNYYNRVYGEFMRVWISGEETLIISKSSSMFHIMKHNHYSSRFGSKLGLQCIGMHEKGIIFNNNPELWKTTRPFFMKALSGPGLVRMVTVCAESLKTHLLLFTPASVN
Specifications
AAH22896 | |
cytochrome P450, family 19, subfamily A, polypeptide 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CYP19A1 | |
GST |
1588 | |
Wheat germ expression system | |
10 μg | |
ARO, ARO1, CPV1, CYAR, CYP19, MGC104309, P-450AROM | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |