missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CYP19A1 (Human) Recombinant Protein

Human CYP19A1 full-length ORF ( AAH22896, 1 a.a. - 181 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00001588-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16136984

  • 6150.00 SEK / 25µg
Estimated Shipment: 19-09-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MVLEMLNPIHYNITSIVPEAMPAATMPVLLLTGLFLLVWNYEGTSSIPGPGYCMGIGPLISHGRFLWMGIGSACNYYNRVYGEFMRVWISGEETLIISKSSSMFHIMKHNHYSSRFGSKLGLQCIGMHEKGIIFNNNPELWKTTRPFFMKALSGPGLVRMVTVCAESLKTHLLLFTPASVN
Specifications

Specifications

AAH22896
cytochrome P450, family 19, subfamily A, polypeptide 1
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CYP19A1
GST
1588
Wheat germ expression system
25 μg
ARO, ARO1, CPV1, CYAR, CYP19, MGC104309, P-450AROM
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates
Special Offers

Special Offers