Learn More
CYP20A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16702-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
CYP20A1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases that catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. The protein lacks one amino acid of the conserved heme binding site. It also lacks the conserved I-helix motif AGX(D, E)T, suggesting that its substrate may carry its own oxygen.Specifications
CYP20A1 | |
Polyclonal | |
Unconjugated | |
CYP20A1 | |
CYP M, CYP20A1, Cytochrome P450 20A1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
316435, 57404, 77951 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6P7D4, Q6UW02, Q8BKE6 | |
Cyp20a1 | |
CYP20A1 Fusion Protein Ag10099 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |