missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP2C19 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55453-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
CYP2C19 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is known to metabolize many xenobiotics, including the anticonvulsive drug mephenytoin, omeprazole, diazepam and some barbiturates. Polymorphism within its gene is associated with variable ability to metabolize mephenytoin, known as the poor metabolizer and extensive metabolizer phenotypes.Specifications
CYP2C19 | |
Polyclonal | |
Unconjugated | |
CYP2C19 | |
(R) limonene 6 monooxygenase, (S) limonene 6 monooxygenase, (S) limonene 7 monooxygenase, CPCJ, CYP2C, CYP2C19, CYPIIC17, CYPIIC19, Cytochrome P450 11A, Cytochrome P450 254C, Cytochrome P450 2C19, Mephenytoin 4 hydroxylase, P450C2C, P450IIC19 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1557 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P33261 | |
CYP2C19 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |