missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP39A1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16062-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum protein is involved in the conversion of cholesterol to bile acids. Its substrates include the oxysterols 25-hydroxycholesterol, 27-hydroxycholesterol and 24-hydroxycholesterol. Alternate splicing results in multiple transcript variants.Specifications
CYP39A1 | |
Polyclonal | |
Unconjugated | |
CYP39A1 | |
CYP39A1, Cytochrome P450 39A1, hCYP39A1, Oxysterol 7 alpha hydroxylase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51302 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NYL5 | |
CYP39A1 | |
CYP39A1 Fusion Protein Ag8896 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |