missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP3A7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55428-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene, CYP3A7, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme hydroxylates testosterone and dehydroepiandrosterone 3-sulphate, which is involved in the formation of estriol during pregcy. The enzyme also metabolizes some drugs such as aflatoxin B1. This gene is part of a cluster of cytochrome P450 genes on chromosome 7q21.1. Transcript variants have been described, but it is not known whether these transcripts are normally produced.Specifications
CYP3A7 | |
Polyclonal | |
Unconjugated | |
CYP3A7 | |
CP37, CYP3A7, CYPIIIA7, Cytochrome P450 3A7, Cytochrome P450 HFLA, P450 HFLA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1551 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P24462 | |
CYP3A7 | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |