missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP4B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11771-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum. In rodents, the homologous protein has been shown to metabolize certain carcinogens; however, the specific function of the human protein has not been determined. Two transcript variants encoding slightly different isoforms have been found for this gene.Specifications
CYP4B1 | |
Polyclonal | |
Unconjugated | |
CYP4B1 | |
CYP4B1, CYPIVB1, Cytochrome P450 4B1, Cytochrome P450 HP, P 450HP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13120, 1580, 24307 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P13584, P15129, Q64462 | |
Cyp4b1 | |
CYP4B1 Fusion Protein Ag2416 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |