Läs mer
Cyproheptadine beta-N-Oxide, Mikromol™
Produktkod. 30461751
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30461751 4 x 25 mg
30050060 25 mg
30051919 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30461751 Leverantör Mikromol Leverantörsnummer MM0609.0600254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Cyproheptadine beta-N-Oxide, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Cyproheptadine beta-N-Oxide
CAS 60251-34-5
Analyt- eller komponentnamn Cyproheptadine beta-N-Oxide
Frakt skick Room Temperature
Molekylformel C21 H21 N O
Certifikat och efterlevnad ISO 9001
Synonym Cyproheptadine beta-N-Oxide
InChI formel InChI=1S/C21H21NO/c1-22(23)14-12-18(13-15-22)21-19-8-4-2-6-16(19)10-11-17-7-3-5-9-20(17)21/h2-11H,12-15H2,1H3
Lösningstyp Neat
LEDER C[N+]1([O-])CCC(=C2c3ccccc3C=Cc4ccccc24)CC1
IUPAC-namn 4-(dibenzo[1,2-a:2',1'-d][7]annulen-11-ylidene)-1-methyl-1-oxidopiperidin-1-ium
Molekylvikt (g/mol) 303.4
Kvantitet 4 x 25 mg
Formel vikt 303.1623 g/mol