Läs mer
Cytarabine 5’-Monophosphate, TRC
Produktkod. 30367575
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30367575 250 mg
1 options
Produktkod. 30367575 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC998105250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Cytarabine 5'-Monophosphate
CAS 7075-11-8
Molekylformel C9 H14 N3 O8 P
Kvantitet 250 mg
Synonym 2(1H)-Pyrimidinone, 4-amino-1-(5-O-phosphono-β-D-arabinofuranosyl)-, Cytosine, 1-β-D-arabinofuranosyl-, 5'-(dihydrogen phosphate) (7CI,8CI), 4-Amino-1-(5-O-phosphono-β-D-arabinofuranosyl)-2(1H)-pyrimidinone, 1-β-D-Arabinofuranosylcytosine 5'-monophosphate, 1-β-D-Arabinofuranosylcytosine 5'-phosphate, Aracytidine 5'-monophosphate, Aracytidine 5'-phosphate, Cytosine arabinoside 5'-phosphate, Cytosine arabinoside monophosphate, Cytosine β-D-arabinoside 5'-monophosphate, Cytosine-β-D-arabinofuranoside-5'-monophosphoric acid, NSC 99445, Cytarabine 5'-monophosphate
InChI formel InChI=1S/C9H14N3O8P/c10-5-1-2-12(9(15)11-5)8-7(14)6(13)4(20-8)3-19-21(16,17)18/h1-2,4,6-8,13-14H,3H2,(H2,10,11,15)(H2,16,17,18)/t4-,6-,7+,8-/m1/s1
LEDER NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](COP(=O)(O)O)[C@@H](O)[C@@H]2O
IUPAC-namn [(2R,3S,4S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
Molekylvikt (g/mol) 323.2
Formel vikt 323.0519