Läs mer
Cytidine 3'(2’)-Monophosphate, TRC
Produktkod. 30352543
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30352543 25 mg -
30354296 10 mg -
30363667 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30352543 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC99831025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 84-52-6
Molekylvikt (g/mol) 323.2
IUPAC-namn [(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl] dihydrogen phosphate
LEDER NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](OP(=O)(O)O)[C@H]2O
Specifikationer
Kemiskt namn eller material Cytidine-3'-monophosphate
Molekylformel C9 H14 N3 O8 P
Kvantitet 25 mg
InChI formel InChI=1S/C9H14N3O8P/c10-5-1-2-12(9(15)11-5)8-6(14)7(4(3-13)19-8)20-21(16,17)18/h1-2,4,6-8,13-14H,3H2,(H2,10,11,15)(H2,16,17,18)/t4-,6-,7-,8-/m1/s1
Formel vikt 323.0519