missing translation for 'onlineSavingsMsg'
Läs mer

Cytidine 3'(2’)-Monophosphate, TRC

Produktkod. 30354296
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
2.5 mg
10 mg
25 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
30354296 10 mg -
30363667 2.5 mg 2.5mg
30352543 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30354296 Supplier Toronto Research Chemicals Supplier No. TRCC99831010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molekylvikt (g/mol) 323.2
Synonym 3'(2’)-Cytidylic Acid, 3'(2’)-CMP, 3'(2’)-Cytidinephosphoric Acid, 3'(2’)-Cytosylic Acid, Ribocytidine 3'(2’)-Monophosphate
LEDER NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](OP(=O)(O)O)[C@H]2O

Specifications

Kemiskt namn eller material Cytidine 3'(2’)-Monophosphate
Molekylformel C9H14N3O8P
Kvantitet 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.