Learn More
Thermo Scientific Chemicals Cytidine-5'-diphosphate disodium salt, 95%
Product Code. 15496269
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
15496269 1 g 1g
1 options
Product Code. 15496269 Supplier Thermo Scientific Chemicals Supplier No. J64234.03
 Restricted Item
I lager
Add to basket
CAS: 54394-90-0 | C9H13N3Na2O11P2 | 447.14 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 54394-90-0
Color White
Molecular Formula C9H13N3Na2O11P2
MDL Number MFCD00084682
Quantity 1 g
Synonym CDP disodium salt
InChI Key ZGDYAAWYYNVXEB-WFIJOQBCSA-L
SMILES [Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]1O
IUPAC Name disodium;(2R,3R,4S,5S)-2-(4-amino-2-oxopyrimidin-1-yl)-5-[[hydroxy(phosphonooxy)phosphoryl]oxymethyl]oxolane-3,4-diolate
Molecular Weight (g/mol) 447.14
PubChem CID 131632379
Formula Weight 447.1
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Cytidine-5'-diphosphate disodium salt