missing translation for 'onlineSavingsMsg'
Läs mer

Cytidine-5'-diphosphate trisodium salt, Thermo Scientific™

Produktkod. 15456339
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
500 mg
1 g
Förpackningsstorlek:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15456339 500 mg 500mg
15446339 100 mg 100mg
15426339 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15456339 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer J64350.ME
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Cytidine-5′-diphosphate trisodium salt has been used to produce a vaccinia virus that induces ribonucleotide reductase in primate cells.

Solubility
Freely soluble in water, insoluble in ethanol, chloroform and ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 34393-59-4
Molekylformel C9H12N3Na3O11P2
Molekylvikt (g/mol) 469.12
MDL-nummer MFCD00065201
InChI-nyckel JENUKVZGXGULDX-LLWADOMFSA-K
Synonym cytidine 5'-diphosphate trisodium salt
PubChem CID 131674171
IUPAC-namn [(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate;molecular hydrogen;sodium
LEDER [Na+].[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]1O

Specifikationer

Kemiskt namn eller material Cytidine-5'-diphosphate trisodium salt
Färg White
Kvantitet 500 mg
Löslighetsinformation Freely soluble in water,insoluble in ethanol,chloroform and ether.
Formel vikt 469.12
Fysisk form Crystalline Powder
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.