Läs mer
Cytidine 5'-Phosphoromorpholidate N,N'-Dicyclohexyl-4-morpholinecarboximidamide, TRC
Produktkod. 30133867
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133867 1 g
30010112 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133867 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9983801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 76742-18-2
Molekylformel C13 H20 N4 O8 P . C17 H32 N3 O
Molekylvikt (g/mol) 685.75
Synonym Cytidine 5'-(Hydrogen 4-Morpholinylphosphonate) N,N'-Dicyclohexyl-4-morpholinecarboximidamide
IUPAC-namn [(2R,3S,4R,5R)-5-(4-amino-2-oxo-pyrimidin-1-yl)-3,4-dihydroxy-tetrahydrofuran-2-yl]methoxy-morpholino-phosphinate;cyclohexyl-[(E)-N-cyclohexyl-C-morpholino-carbonimidoyl]ammonium
LEDER NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](COP(=O)([O-])N3CCOCC3)[C@@H](O)[C@H]2O.C4CCC(CC4)[NH2+]\C(=N/C5CCCCC5)\N6CCOCC6
Specifikationer
Kemiskt namn eller material Cytidine 5'-Phosphoromorpholidate N,N'-Dicyclohexyl-4-morpholinecarboximidamide
Kvantitet 1 g
InChI formel InChI=1S/C17H31N3O.C13H21N4O8P/c1-3-7-15(8-4-1)18-17(20-11-13-21-14-12-20)19-16-9-5-2-6-10-16;14-9-1-2-17(13(20)15-9)12-11(19)10(18)8(25-12)7-24-26(21,22)16-3-5-23-6-4-16/h15-16H,1-14H2,(H,18,19);1-2,8,10-12,18-19H,3-7H2,(H,21,22)(H2,14,15,20)/t;8-,10-,11-,12-/m.1/s1
Formel vikt 685.356