Learn More
Hettich™ Cyto Chamber
Designed for use in cytology applications
Brand:  Hettich™ 1667
Additional Details : Weight : 0.01000kg
Product Code. 12083884
  • 7930.00 SEK / Pack of 100
Estimated Shipment
11-12-2019
Add to basket
Description
Description
  • Cyto Chamber
  • For slide carrier no. 1662 and 1670
  • Fits carriers 1660 and 1452 and two-tier frame 5280
  • One-funnel chambers:
  • 1mL chambers for small quantities of samples which are low in cells, such as cerebrospinal fluid
  • 2/4mL chambers for samples rich in cells such as pleural effusions, ascites and BAL
  • 8mL chambers for high volume samples such as urine
  • Multi-funnel chambers:
  • If a number of sediments are required from a single sample (for example for tests with different antibodies) then the multi-funnel chambers may be used
  • If the 4-place chambers are used in combination with the double slide carrier then up to 8 individual sediments can be prepared per microscope slide
Specifications
Specifications
3 x 8.7mm
Slide Carriers 1662 and 1670
3
3 x 2mL
Cyto Chamber