Learn More
MedChemExpress Cytochalasin B
Product Code. 16491649
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity Unit Size
16491649 1 mg 1mg
1 options
Product Code. 16491649 Supplier MedChemExpress Supplier No. HY16928
 Restricted Item
Beräknad leverans: 10-12-2026
MedChemExpress Cytochalasin B is a cell-permeable mycotoxin binding to the barbed end of actin filaments, disrupting the formation of actin polymers, with Kd value of 1.4-2.2 nM for F-actin.
Specifications
Content And Storage Powder -20°C 3 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 479.61 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 14930-96-2
Product Type Natural Products
Purity 98.0%
Solubility DMSO : 83.33 mg/mL (173.75 mM∣ultrasonic and warming and heat to 60°C) ∣Ethanol : 25 mg/mL (52.13 mM∣Need ultrasonic)
Synonym Phomin
Target Arp2/3 Complex
Quantity 1 mg